siRNA Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004419.3 | NP_004410.3 | 3 | 923 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUSP5| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004419.3 | NP_004410.3 | 3 | 923 | ||