siRNA Details
| Gene Symbol: EFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004429.4 | NP_004420.1 | 5 | 1578 | ||
| GenBank mRNA | BC016649.1 | - | 5 | 1578 | 3867060 | |
| BC052979.1 | - | 5 | 1597 | 6164440 | ||
| L37361.1 | - | 5 | 1324 | |||
| U09303.1 | - | 5 | 1499 | |||
| U09304.1 | - | 5 | 1105 | |||

EFNB1
Genomic Map