siRNA Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300814.1 | NP_001287743.1 | 9 | 1009 | ||
| NM_173352.3 | NP_775487.2 | 9 | 1391 | |||
| XM_011538010.1 | XP_011536312.1 | 8 | 1295 | |||
| GenBank mRNA | AK096419.1 | - | 9 | 1391 | ||
| AK290901.1 | - | 9 | 1391 | |||
| AK298636.1 | - | 10 | 1317 | |||
| BX647095.1 | - | 10 | 1009 | |||

KRT78
Genomic Map