siRNA Details
| Gene Symbol: FAM71C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153364.3 | NP_699195.1 | 2 | 1075 | ||
| GenBank mRNA | BC031221.1 | - | 2 | 1072 | 5272209 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM71C| Gene Symbol: FAM71C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153364.3 | NP_699195.1 | 2 | 1075 | ||
| GenBank mRNA | BC031221.1 | - | 2 | 1072 | 5272209 | |