siRNA Details
| Gene Symbol: CT62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102658.1 | NP_001096128.1 | 3 | 526 | ||
| XM_006720429.3 | XP_006720492.1 | 3 | 786 | |||
| GenBank mRNA | AL080151.1 | - | 3 | 797 | ||
| BC039359.1 | - | 3 | 522 | |||
| BC048128.1 | - | 3 | 488 | |||

CT62
Genomic Map