siRNA Details
| Gene Symbol: GRAMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012642.2 | NP_001012660.1 | 1, 2 | 63 | ||
| XM_011521331.2 | XP_011519633.1 | 1, 2 | 63 | |||
| XM_017021997.1 | XP_016877486.1 | 1, 2 | 82 | |||
| GenBank mRNA | AK095072.1 | - | 1, 2 | 53 | ||
| BC038451.1 | - | 1, 2 | 37 | |||
| DA854662.1 | - | 1, 2 | 63 | |||

GRAMD2
Genomic Map