siRNA Details
| Gene Symbol: GRAMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012642.2 | NP_001012660.1 | 8 | 586 | ||
| XM_011521327.2 | XP_011519629.1 | 8 | 695 | |||
| XM_011521328.2 | XP_011519630.1 | 8 | 1046 | |||
| XM_011521329.2 | XP_011519631.1 | 8 | 563 | |||
| XM_011521330.2 | XP_011519632.1 | 8 | 647 | |||
| XM_011521332.2 | XP_011519634.1 | 9 | 770 | |||
| XM_011521333.2 | XP_011519635.1 | 9 | 787 | |||
| XM_017021997.1 | XP_016877486.1 | 8 | 730 | |||
| XM_017021998.1 | XP_016877487.1 | 8 | 954 | |||
| GenBank mRNA | AK095072.1 | - | 8 | 576 | ||
| BC038451.1 | - | 8 | 560 | |||

GRAMD2
Genomic Map