siRNA Details
| Gene Symbol: TMEM61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182532.2 | NP_872338.1 | 2 | 508 | ||
| XM_005270586.4 | XP_005270643.1 | 2 | 146 | |||
| XM_011540911.2 | XP_011539213.1 | 3 | 260 | |||
| XM_011540912.2 | XP_011539214.1 | 2 | 1036 | |||
| GenBank mRNA | BC029775.1 | - | 2 | 380 | 5186268 | |
| BG430102.1 | - | 2 | 508 | |||

TMEM61
Genomic Map