siRNA Details
| Gene Symbol: KRTCAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173852.3 | NP_776251.1 | 5 | 487 | ||
| GenBank mRNA | AK024167.1 | - | 4 | 3204 | ||
| AK311907.1 | - | 5 | 461 | |||
| AY157577.1 | - | 4 | 372 | |||
| BC029806.1 | - | 5 | 474 | 5189880 | ||
| BC048205.1 | - | 5 | 413 | |||
| BC057233.1 | - | 5 | 496 | 6463372 | ||
| BX647396.1 | - | 4 | 2811 | |||

KRTCAP2
Genomic Map