siRNA Details
| Gene Symbol: RNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017981.1 | NP_001017981.1 | 3 | 459 | ||
| GenBank mRNA | AK293904.1 | - | 2 | 301 | ||
| BM476400.1 | - | 3 | 271 | |||
| BX494669.1 | - | 4 | 411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF215| Gene Symbol: RNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017981.1 | NP_001017981.1 | 3 | 459 | ||
| GenBank mRNA | AK293904.1 | - | 2 | 301 | ||
| BM476400.1 | - | 3 | 271 | |||
| BX494669.1 | - | 4 | 411 | |||