siRNA Details
| Gene Symbol: ANKRD23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144994.7 | NP_659431.5 | 8 | 803 | ||
| GenBank mRNA | AF492401.1 | - | 8 | 779 | ||
| AJ315766.1 | - | 7 | 747 | |||
| AK092564.1 | - | 7 | 677 | |||
| AY196212.1 | - | 8 | 787 | |||
| BC107056.2 | - | 8 | 798 | 40005436 | ||

ANKRD23
Genomic Map