siRNA Details
| Gene Symbol: STRA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271007.1 | NP_001257936.1 | 2, 3 | 127 | ||
| NM_144998.3 | NP_659435.2 | 2, 3 | 127 | |||
| XM_017024329.1 | XP_016879818.1 | 2, 3 | 148 | |||
| GenBank mRNA | BC009571.1 | - | 2, 3 | 96 | 4333630 | |
| BC011610.1 | - | 2, 3 | 78 | |||
| BI837753.1 | - | 2, 3 | 95 | |||
| U95006.1 | - | 2, 3 | 82 | |||
| U95007.1 | - | 2, 3 | 82 | |||

STRA13
Genomic Map