siRNA Details
| Gene Symbol: EMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001313905.1 | NP_001300834.1 | 4 | 354 | ||
| NM_001425.2 | NP_001416.1 | 4 | 538 | |||
| XM_011526605.2 | XP_011524907.1 | 5 | 469 | |||
| GenBank mRNA | AL541088.3 | - | 4 | 538 | ||
| BC009718.1 | - | 4 | 345 | 3891050 | ||
| BT007124.1 | - | 3 | 284 | |||
| CR456871.1 | - | 3 | 284 | |||
| CR541955.1 | - | 3 | 284 | |||
| U52101.1 | - | 4 | 333 | |||
| U87947.1 | - | 4 | 525 | |||

EMP3
Genomic Map