siRNA Details
| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004097.2 | NP_004088.2 | 1 | 512 | ||
| XM_005264203.3 | XP_005264260.1 | 1 | 521 | |||
| GenBank mRNA | BC045762.1 | - | 1 | 512 | 5260039 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMX1| Gene Symbol: EMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004097.2 | NP_004088.2 | 1 | 512 | ||
| XM_005264203.3 | XP_005264260.1 | 1 | 521 | |||
| GenBank mRNA | BC045762.1 | - | 1 | 512 | 5260039 | |