siRNA Details
| Gene Symbol: CMYA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153610.4 | NP_705838.3 | 2 | 4323 | ||
| XM_017009212.1 | XP_016864701.1 | 2 | 4191 | |||
| GenBank mRNA | AK096453.1 | - | 1 | 410 | ||
| AL832347.1 | - | 2 | 4125 | |||
| BX647452.1 | - | 1 | 469 | |||

CMYA5
Genomic Map