siRNA Details
| Gene Symbol: PAXIP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028090.1 | 1 | 506 | |||
| GenBank mRNA | AL713756.1 | - | 1 | 457 | ||
| BC035068.1 | - | 1 | 495 | |||
| BC048116.1 | - | 1 | 476 | |||
| BI545940.1 | - | 1 | 493 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAXIP1-AS1| Gene Symbol: PAXIP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028090.1 | 1 | 506 | |||
| GenBank mRNA | AL713756.1 | - | 1 | 457 | ||
| BC035068.1 | - | 1 | 495 | |||
| BC048116.1 | - | 1 | 476 | |||
| BI545940.1 | - | 1 | 493 | |||