siRNA Details
| Gene Symbol: PAXIP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028090.1 | 1 | 585 | |||
| GenBank mRNA | AK098428.1 | - | 1 | 65 | ||
| AL713756.1 | - | 1 | 536 | |||
| BC035068.1 | - | 1 | 574 | |||
| BC048116.1 | - | 1 | 555 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAXIP1-AS1| Gene Symbol: PAXIP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028090.1 | 1 | 585 | |||
| GenBank mRNA | AK098428.1 | - | 1 | 65 | ||
| AL713756.1 | - | 1 | 536 | |||
| BC035068.1 | - | 1 | 574 | |||
| BC048116.1 | - | 1 | 555 | |||