siRNA Details
| Gene Symbol: CT83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017978.3 | NP_001017978.1 | 2 | 416 | ||
| GenBank mRNA | AK000618.1 | - | 2 | 386 | ||
| AK026566.1 | - | 2 | 386 | |||
| BC062223.1 | - | 2 | 363 | 6101827 | ||
| HY170569.1 | - | 2 | 417 | |||

CT83
Genomic Map