siRNA Details
| Gene Symbol: EPHB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004444.4 | NP_004435.3 | 10 | 2225 | ||
| XM_017011816.1 | XP_016867305.1 | 10 | 1882 | |||
| GenBank mRNA | AB209644.1 | - | 9 | 2935 | ||
| AK301224.1 | - | 5 | 532 | |||
| AY056048.1 | - | 10 | 1819 | |||
| BC004264.1 | - | 7 | 1310 | |||
| BC052804.1 | - | 10 | 1902 | 6584384 | ||
| U07695.1 | - | 10 | 1819 | |||

EPHB4
Genomic Map