siRNA Details
| Gene Symbol: ESRRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282450.1 | NP_001269379.1 | 7 | 1420 | ||
| NM_001282451.1 | NP_001269380.1 | 7 | 1404 | |||
| NM_004451.4 | NP_004442.3 | 7 | 1407 | |||
| XM_017017313.1 | XP_016872802.1 | 6 | 1911 | |||
| GenBank mRNA | BC007915.2 | - | 6 | 877 | ||
| BC011528.1 | - | 1 | 83 | |||
| BC033701.1 | - | 5 | 816 | |||
| BC063795.1 | - | 7 | 1357 | 6527166 | ||
| BC092470.1 | - | 7 | 1420 | 30336569 | ||
| BC093720.1 | - | 6 | 1208 | 7939565 | ||
| BC093722.1 | - | 6 | 1208 | 7939567 | ||
| HQ692867.1 | - | 6 | 1186 | |||
| L38487.1 | - | 7 | 1360 | |||
| X51416.1 | - | 1543 | ||||

ESRRA
Genomic Map