siRNA Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306090.1 | NP_001293019.1 | 7 | 1105 | ||
| NM_001306092.1 | NP_001293021.1 | 7 | 1105 | |||
| NM_153717.2 | NP_714928.1 | 7 | 1105 | |||
| XM_006713865.3 | XP_006713928.1 | 7 | 1102 | |||
| XM_006713866.3 | XP_006713929.1 | 7 | 1102 | |||
| XM_011513419.2 | XP_011511721.1 | 7 | 1099 | |||
| XM_017007883.1 | XP_016863372.1 | 7 | 1099 | |||
| GenBank mRNA | AF216184.1 | - | 7 | 1105 | ||
| AF216185.1 | - | 7 | 1105 | |||
| AF239742.1 | - | 7 | 921 | |||
| AK025394.1 | - | 2 | 234 | |||
| BC085608.1 | - | 7 | 1087 | 30523373 | ||

EVC
Genomic Map