siRNA Details
| Gene Symbol: EVX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304519.1 | NP_001291448.1 | 1 | 552 | ||
| NM_001304520.1 | NP_001291449.1 | 1 | 552 | |||
| NM_001989.4 | NP_001980.1 | 1 | 552 | |||
| GenBank mRNA | AK299188.1 | - | 1 | 552 | ||
| AK314155.1 | - | 1 | 397 | |||
| X60655.1 | - | 1 | 294 | |||

EVX1
Genomic Map