siRNA Details
| Gene Symbol: FABP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040442.1 | NP_001035532.1 | 4, 5 | 453 | ||
| NM_001130958.1 | NP_001124430.1 | 5, 6 | 533 | |||
| NM_001445.2 | NP_001436.1 | 2, 3 | 368 | |||
| GenBank mRNA | BC022489.1 | - | 3, 4 | 338 | 4792594 | |
| DQ132786.1 | - | 5, 6 | 534 | |||
| U19869.1 | - | 2, 3 | 289 | |||
| X90908.1 | - | 2, 3 | 368 | |||

FABP6
Genomic Map