siRNA Details
| Gene Symbol: ACSL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004457.3 | NP_004448.2 | 14 | 2166 | ||
| NM_203372.1 | NP_976251.1 | 13 | 2059 | |||
| XM_017003584.1 | XP_016859073.1 | 12 | 1978 | |||
| GenBank mRNA | AB061712.1 | - | 14 | 2134 | ||
| AF116690.1 | - | 2 | 255 | |||
| AK001471.1 | - | 9 | 1085 | |||
| AK314621.1 | - | 14 | 2144 | |||
| BC032144.2 | - | 1 | 47 | |||
| BC041692.1 | - | 14 | 2158 | 5226905 | ||
| D89053.1 | - | 12 | 1857 | |||

ACSL3
Genomic Map