siRNA Details
| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 3, 4 | 705 | |||
| GenBank mRNA | AK096758.1 | - | 3, 4 | 407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM254-AS1| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 3, 4 | 705 | |||
| GenBank mRNA | AK096758.1 | - | 3, 4 | 407 | ||