siRNA Details
| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 4, 5 | 900 | |||
| NR_027429.1 | 3, 4 | 813 | ||||
| GenBank mRNA | AK021673.1 | - | 3, 4 | 532 | ||
| AK096758.1 | - | 4, 5 | 602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM254-AS1| Gene Symbol: TMEM254-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027428.1 | 4, 5 | 900 | |||
| NR_027429.1 | 3, 4 | 813 | ||||
| GenBank mRNA | AK021673.1 | - | 3, 4 | 532 | ||
| AK096758.1 | - | 4, 5 | 602 | |||