siRNA Details
| Gene Symbol: ZCCHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153367.3 | NP_699198.2 | 4 | 877 | ||
| XM_011539452.2 | XP_011537754.1 | 4 | 828 | |||
| GenBank mRNA | AK293361.1 | - | 4 | 516 | ||
| AK303734.1 | - | 4 | 508 | |||
| AK308876.1 | - | 5 | 808 | |||
| BC028617.1 | - | 4 | 879 | 4821347 | ||
| BC039249.1 | - | 4 | 872 | |||
| BC053564.1 | - | 4 | 776 | 6193781 | ||

ZCCHC24
Genomic Map