siRNA Details
| Gene Symbol: FAM124A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242312.1 | NP_001229241.1 | 4 | 1738 | ||
| NM_145019.3 | NP_659456.3 | 5 | 1846 | |||
| XM_011534978.2 | XP_011533280.1 | 6 | 1870 | |||
| XM_017020419.1 | XP_016875908.1 | 5 | 1805 | |||
| GenBank mRNA | AK055269.1 | - | 4 | 1705 | ||
| AK056643.1 | - | 1 | 432 | |||
| AK096364.1 | - | 5 | 1809 | |||
| AY358177.1 | - | 1 | 740 | |||
| BC051771.1 | - | 4 | 1653 | 5722982 | ||

FAM124A
Genomic Map