siRNA Details
| Gene Symbol: DOK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152721.5 | NP_689934.2 | 7 | 1030 | ||
| XM_011525875.2 | XP_011524177.1 | 7 | 1051 | |||
| XM_017025610.1 | XP_016881099.1 | 5 | 3091 | |||
| XM_017025611.1 | XP_016881100.1 | 5 | 1605 | |||
| GenBank mRNA | AK057795.1 | - | 6 | 739 | ||
| AY599248.1 | - | 7 | 1028 | |||
| BC008583.1 | - | 5 | 612 | |||
| BC019045.2 | - | 5 | 655 | |||
| BC096744.1 | - | 7 | 1023 | 5260167 | ||

DOK6
Genomic Map