siRNA Details
| Gene Symbol: RASGEF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282862.1 | NP_001269791.1 | 7 | 918 | ||
| NM_145313.3 | NP_660356.2 | 7 | 890 | |||
| XM_005271809.3 | XP_005271866.1 | 6 | 768 | |||
| XM_011539500.2 | XP_011537802.1 | 6 | 878 | |||
| GenBank mRNA | AK095136.1 | - | 7 | 918 | ||
| AK127432.1 | - | 7 | 3316 | |||
| BC022548.1 | - | 7 | 890 | 4814750 | ||

RASGEF1A
Genomic Map