siRNA Details
Gene Symbol: FAM162B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085480.2 | NP_001078949.1 | 2, 3 | 427 | ||
GenBank mRNA | BC038997.1 | - | 2, 3 | 426 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM162B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085480.2 | NP_001078949.1 | 2, 3 | 427 | ||
GenBank mRNA | BC038997.1 | - | 2, 3 | 426 |