siRNA Details
Gene Symbol: SMIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 1151 | ||
GenBank mRNA | AK299804.1 | - | 1 | 360 | ||
BC032356.1 | - | 2 | 1151 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 1151 | ||
GenBank mRNA | AK299804.1 | - | 1 | 360 | ||
BC032356.1 | - | 2 | 1151 |