siRNA Details
| Gene Symbol: SDK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079653.1 | NP_001073121.1 | 10, 11 | 1488 | ||
| NM_152744.3 | NP_689957.3 | 36, 37 | 5343 | |||
| XM_011515188.1 | XP_011513490.1 | 30, 31 | 4184 | |||
| XM_011515189.1 | XP_011513491.1 | 30, 31 | 4181 | |||
| XM_011515190.1 | XP_011513492.1 | 30, 31 | 4184 | |||
| XM_017011837.1 | XP_016867326.1 | 30, 31 | 4184 | |||
| GenBank mRNA | AK074077.1 | - | 3, 4 | 283 | ||
| AK074083.1 | - | 21, 22 | 2979 | |||
| AL833354.1 | - | 10, 11 | 1485 | |||
| AY310398.1 | - | 36, 37 | 5343 | |||

SDK1
Genomic Map