siRNA Details
| Gene Symbol: FOXK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037165.1 | NP_001032242.1 | 7, 8 | 1715 | ||
| XM_011515191.2 | XP_011513493.1 | 9, 10 | 1386 | |||
| GenBank mRNA | AK122663.1 | - | 8, 9 | 1205 | ||
| AK302243.1 | - | 7, 8 | 1354 | |||
| BC038434.1 | - | 10, 11 | 1459 | |||

FOXK1
Genomic Map