siRNA Details
| Gene Symbol: THSD7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015204.2 | NP_056019.1 | 6 | 1960 | ||
| XM_006715659.1 | XP_006715722.1 | 6 | 1959 | |||
| XM_006715660.1 | XP_006715723.1 | 6 | 1959 | |||
| XM_006715662.1 | XP_006715725.1 | 6 | 1959 | |||
| XM_011515193.2 | XP_011513495.1 | 6 | 4284 | |||
| XM_011515194.1 | XP_011513496.1 | 6 | 1725 | |||
| GenBank mRNA | AB023177.2 | - | 5 | 1244 | ||
| AK092252.1 | - | 6 | 1960 | |||

THSD7A
Genomic Map