siRNA Details
| Gene Symbol: PRR15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175887.2 | NP_787083.1 | 2 | 978 | ||
| XM_005249653.3 | XP_005249710.1 | 2 | 1781 | |||
| XM_011515198.1 | XP_011513500.1 | 2 | 1776 | |||
| XM_011515199.1 | XP_011513501.1 | 2 | 690 | |||
| XM_017011842.1 | XP_016867331.1 | 2 | 988 | |||
| GenBank mRNA | BC018144.1 | - | 2 | 875 | ||
| BC029131.1 | - | 2 | 978 | 5189352 | ||
| FJ857942.1 | - | 1 | 298 | |||

PRR15
Genomic Map