siRNA Details
| Gene Symbol: LRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317721.1 | NP_001304650.1 | 2 | 368 | ||
| NM_152892.2 | NP_690852.1 | 2 | 350 | |||
| XM_017011845.1 | XP_016867334.1 | 2 | 251 | |||
| GenBank mRNA | AB209379.1 | - | 2 | 228 | ||
| AK290967.1 | - | 2 | 249 | |||
| AK313771.1 | - | 2 | 233 | |||
| AL133057.1 | - | 2 | 300 | |||
| BC009436.1 | - | 1 | 41 | |||
| BC030547.1 | - | 2 | 290 | 5170588 | ||

LRWD1
Genomic Map