siRNA Details
| Gene Symbol: ZNRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147128.3 | NP_667339.1 | 2, 3 | 1612 | ||
| GenBank mRNA | AF513707.1 | - | 2, 3 | 1612 | ||
| AF527533.1 | - | 2, 3 | 939 | |||
| AK025240.1 | - | 1, 2 | 54 | |||
| AY054122.1 | - | 3, 4 | 376 | |||

ZNRF2
Genomic Map