siRNA Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320666.1 | NP_001307595.1 | 4, 5 | 752 | ||
| NM_001320667.1 | NP_001307596.1 | 4, 5 | 658 | |||
| NM_001320668.1 | NP_001307597.1 | 4, 5 | 528 | |||
| NM_001320669.1 | NP_001307598.1 | 4, 5 | 466 | |||
| NM_014898.2 | NP_055713.1 | 4, 5 | 699 | |||
| GenBank mRNA | AB023178.1 | - | 4, 5 | 699 | ||
| BC041087.1 | - | 4, 5 | 728 | 5579247 | ||
| BC050542.1 | - | 4, 5 | 657 | |||
| GU388430.1 | - | 4, 5 | 528 | |||

ZFP30
Genomic Map