siRNA Details
| Gene Symbol: RNF44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014901.4 | NP_055716.1 | 3 | 804 | ||
| XM_005265840.2 | XP_005265897.1 | 3 | 516 | |||
| XM_005265841.4 | XP_005265898.1 | 3 | 761 | |||
| XM_005265842.4 | XP_005265899.1 | 3 | 746 | |||
| XM_005265843.2 | XP_005265900.1 | 3 | 452 | |||
| XM_005265844.3 | XP_005265901.1 | 3 | 557 | |||
| XM_005265845.4 | XP_005265902.1 | 3 | 695 | |||
| XM_006714831.2 | XP_006714894.1 | 3 | 516 | |||
| XM_006714832.2 | XP_006714895.1 | 3 | 516 | |||
| XM_011534466.2 | XP_011532768.1 | 3 | 519 | |||
| XM_011534467.2 | XP_011532769.1 | 3 | 750 | |||
| GenBank mRNA | AB029023.1 | - | 3 | 689 | ||
| AK302385.1 | - | 3 | 557 | |||
| BC039833.1 | - | 3 | 760 | 5535911 | ||
| BC063297.1 | - | 3 | 805 | 30349418 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_134064, NM_001146025, NM_001146026, NM_001146027, AK171943, and others.

RNF44
Genomic Map