siRNA Details
| Gene Symbol: CHSY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014918.4 | NP_055733.2 | 3 | 2379 | ||
| XM_006720435.3 | XP_006720498.1 | 3 | 1661 | |||
| XM_011521364.2 | XP_011519666.1 | 4 | 2002 | |||
| XM_017022011.1 | XP_016877500.1 | 2 | 1435 | |||
| GenBank mRNA | AK075096.1 | - | 1 | 109 | ||
| AK296943.1 | - | 2 | 1156 | |||
| AY358529.1 | - | 3 | 2391 | |||
| BC046247.1 | - | 3 | 2386 | 5751618 | ||

CHSY1
Genomic Map