siRNA Details
| Gene Symbol: MON1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286639.1 | NP_001273568.1 | 5 | 1642 | ||
| NM_001286640.1 | NP_001273569.1 | 4 | 1484 | |||
| NM_014940.3 | NP_055755.1 | 6 | 1969 | |||
| GenBank mRNA | AB020679.1 | - | 6 | 1653 | ||
| AF442486.1 | - | 5 | 1633 | |||
| AK023374.1 | - | 5 | 910 | |||
| AK023896.1 | - | 6 | 1697 | |||
| AK128411.1 | - | 3 | 1511 | |||
| AK293392.1 | - | 4 | 1204 | |||
| AK296467.1 | - | 5 | 1369 | |||
| BC024277.1 | - | 6 | 1949 | 4843859 | ||

MON1B
Genomic Map