siRNA Details
| Gene Symbol: KIAA0907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014949.3 | NP_055764.2 | 14 | 1759 | ||
| GenBank mRNA | AB020714.1 | - | 13 | 3269 | ||
| AY112680.1 | - | 14 | 1690 | |||
| BC062637.1 | - | 14 | 1714 | 5497581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KIAA0907| Gene Symbol: KIAA0907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014949.3 | NP_055764.2 | 14 | 1759 | ||
| GenBank mRNA | AB020714.1 | - | 13 | 3269 | ||
| AY112680.1 | - | 14 | 1690 | |||
| BC062637.1 | - | 14 | 1714 | 5497581 | ||