siRNA Details
| Gene Symbol: ZNF365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199451.2 | NP_955523.1 | 6, 7 | 1543 | ||
| NM_199452.3 | NP_955524.3 | 5, 6 | 824 | |||
| XM_017015938.1 | XP_016871427.1 | 4, 5 | 680 | |||
| XM_017015940.1 | XP_016871429.1 | 4, 5 | 647 | |||
| XM_017015941.1 | XP_016871430.1 | 4, 5 | 830 | |||
| GenBank mRNA | AJ505149.1 | - | 6, 7 | 1505 | ||
| AJ505150.1 | - | 5, 6 | 824 | |||
| BC101085.2 | - | 3, 4 | 378 | |||

ZNF365
Genomic Map