siRNA Details
| Gene Symbol: SBNO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100122.1 | NP_001093592.1 | 12 | 1537 | ||
| NM_014963.2 | NP_055778.2 | 15 | 1869 | |||
| XM_011527804.2 | XP_011526106.1 | 15 | 1842 | |||
| GenBank mRNA | AB023180.1 | - | 15 | 1846 | ||
| AK074102.1 | - | 11 | 1329 | |||
| AK125139.1 | - | 12 | 1484 | |||
| AK296849.1 | - | 12 | 1484 | |||
| AK300981.1 | - | 16 | 1809 | |||
| AK309167.1 | - | 12 | 1484 | |||
| BG332816.1 | - | 1 | 54 | |||
| BQ950129.1 | - | 5 | 628 | |||

SBNO2
Genomic Map