siRNA Details
| Gene Symbol: MMRN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007351.2 | NP_031377.2 | 1, 2 | 699 | ||
| XM_005262856.2 | XP_005262913.1 | 1, 2 | 724 | |||
| XM_017007893.1 | XP_016863382.1 | 2, 3 | 940 | |||
| GenBank mRNA | AK125557.1 | - | 2, 3 | 646 | ||
| AK313566.1 | - | 1, 2 | 724 | |||
| BC063848.1 | - | 1, 2 | 699 | 6180674 | ||
| U27109.1 | - | 1, 2 | 699 | |||

MMRN1
Genomic Map