siRNA Details
| Gene Symbol: ZP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207341.3 | NP_997224.2 | 9, 10 | 1586 | ||
| XM_011544852.2 | XP_011543154.1 | 6, 7 | 1102 | |||
| XM_011544853.2 | XP_011543155.1 | 7, 8 | 965 | |||
| GenBank mRNA | AI208352.1 | - | 3, 4 | 396 | ||
| BC067899.1 | - | 5, 6 | 1185 | 5298273 | ||
| EU794684.1 | - | 9, 10 | 1586 | |||

ZP1
Genomic Map