siRNA Details
| Gene Symbol: ZP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207341.3 | NP_997224.2 | 2 | 235 | ||
| GenBank mRNA | EU794684.1 | - | 2 | 235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZP1| Gene Symbol: ZP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207341.3 | NP_997224.2 | 2 | 235 | ||
| GenBank mRNA | EU794684.1 | - | 2 | 235 | ||