siRNA Details
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 13 | 1854 | ||
NM_014985.3 | NP_055800.2 | 13 | 1854 | |||
XM_006720437.3 | XP_006720500.1 | 13 | 2039 | |||
XM_011521373.2 | XP_011519675.1 | 13 | 2607 | |||
XM_011521374.2 | XP_011519676.1 | 13 | 2606 | |||
XM_011521375.2 | XP_011519677.1 | 13 | 2606 | |||
XM_011521378.2 | XP_011519680.1 | 13 | 2606 | |||
XM_011521379.2 | XP_011519681.1 | 13 | 2605 | |||
XM_017022014.1 | XP_016877503.1 | 6 | 800 | |||
XM_017022016.1 | XP_016877505.1 | 13 | 2609 | |||
GenBank mRNA | AB020719.1 | - | 12 | 1459 | ||
AK303805.1 | - | 13 | 1777 | |||
BC107721.1 | - | 13 | 1858 | |||
BC117182.1 | - | 13 | 1695 | 40125733 | ||
BX648822.1 | - | 13 | 1781 |
