siRNA Details
| Gene Symbol: DAAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270520.1 | NP_001257449.1 | 2 | 224 | ||
| NM_014992.2 | NP_055807.1 | 1 | 118 | |||
| XM_005267430.1 | XP_005267487.1 | 2 | 276 | |||
| XM_005267431.1 | XP_005267488.1 | 2 | 376 | |||
| GenBank mRNA | AB014566.1 | - | 1 | 104 | ||
| AK021890.1 | - | 2 | 168 | |||
| AK291790.1 | - | 2 | 193 | |||
| BC038428.1 | - | 2 | 216 | 4993504 | ||
| BC064999.1 | - | 2 | 207 | 5784628 | ||
| BP199650.1 | - | 2 | 224 | |||
| BQ050640.1 | - | 2 | 206 | |||
| BX247986.1 | - | 2 | 301 | |||
| BX648383.1 | - | 2 | 242 | |||

DAAM1
Genomic Map